accessibility menu, dialog, popup

UserName

CAS RN 6298-37-9

H2NNN

CAS RN 6298-37-9

IUPAC Name: quinoxalin-6-amine
Synonyms: 6-aminoquinoxaline
Molecular Weight (g/mol): 145.17
Molecular Formula: C8H7N3
InChi Key: MSGRFBKVMUKEGZ-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=CC=NC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)