accessibility menu, dialog, popup

UserName

CAS RN 64919-15-9

H3COOCl

CAS RN 64919-15-9

IUPAC Name: 2-chloroethyl dodecanoate
Synonyms: 2-chloroethyl dodecanoate
Molecular Weight (g/mol): 262.82
Molecular Formula: C14H27ClO2
InChi Key: PPRUSMUBWUQYRY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results