CAS RN 6638-60-4
CAS RN 6638-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-7-bromofluorene, tech. 90%
CAS: 6638-60-4 Molecular Formula: C13H10BrN Molecular Weight (g/mol): 260.134 MDL Number: MFCD00001127 InChI Key: RJWYTISHBMNMOZ-UHFFFAOYSA-N Synonym: 2-amino-7-bromofluorene,2-bromo-7-aminofluorene,fluoren-2-amine, 7-bromo,7-bromo-9h-fluoren-2-ylamine,9h-fluoren-2-amine, 7-bromo,7-bromofluorene-2-ylamine,acmc-20a5iw,2-amino-7-bromo-9h-fluorene,9h-fluoren-2-amine,7-bromo,7-bromanyl-9h-fluoren-2-amine PubChem CID: 23126 IUPAC Name: 7-bromo-9H-fluoren-2-amine SMILES: C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br