accessibility menu, dialog, popup

UserName

CAS RN 66389-76-2

CH3CH3H3COONHOOH3C

CAS RN 66389-76-2

IUPAC Name: tert-butyl N-(methoxycarbonyl)carbamate
Synonyms: tert-butyl N-(methoxycarbonyl)carbamate
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: BPECCYFUJCNQOR-UHFFFAOYSA-N
SMILES: COC(=O)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results