accessibility menu, dialog, popup

UserName

CAS RN 71617-10-2

H3COOOCH3CH3(E)

CAS RN 71617-10-2

IUPAC Name: 3-methylbutyl (2E)-3-(4-methoxyphenyl)prop-2-enoate
Synonyms: neo heliopan amiloxate isoamyl P-methoxycinnamate neo heliopan E1000
Molecular Weight (g/mol): 248.32
Molecular Formula: C15H20O3
InChi Key: UBNYRXMKIIGMKK-RMKNXTFCSA-N
SMILES: COC1=CC=C(\C=C\C(=O)OCCC(C)C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results