accessibility menu, dialog, popup

UserName

CAS RN 7488-55-3

SnOSOOO

CAS RN 7488-55-3

IUPAC Name: λ²-tin(2+) sulfate
Synonyms: tin(II) sulfate
Molecular Weight (g/mol): 214.77
Molecular Formula: O4SSn
InChi Key: OBBXFSIWZVFYJR-UHFFFAOYSA-L
SMILES: [Sn++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)