accessibility menu, dialog, popup

UserName

CAS RN 75804-28-3

CH3CH3CH3CH3H2NH2NHClHCl

CAS RN 75804-28-3

IUPAC Name: 2,3-dimethylbutane-2,3-diamine dihydrochloride
Synonyms: 2,3-dimethylbutane-2,3-diamine dihydrochloride
Molecular Weight (g/mol): 189.12
Molecular Formula: C6H18Cl2N2
InChi Key: BRLVXFONMZRJCD-UHFFFAOYSA-N
SMILES: Cl.Cl.CC(C)(N)C(C)(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results