accessibility menu, dialog, popup

UserName

CAS RN 75853-18-8

HOFF

CAS RN 75853-18-8

IUPAC Name: (2,3-difluorophenyl)methanol
Synonyms: 2,3-difluorobenzyl alcohol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: JSFGDUIJQWWBGY-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results