accessibility menu, dialog, popup

UserName

CAS RN 7682-18-0

H3COOCH3NH2HCl

CAS RN 7682-18-0

IUPAC Name: methyl 2-aminobutanoate hydrochloride
Synonyms: methyl 2-aminobutanoate hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: AHAQQEGUPULIOZ-UHFFFAOYNA-N
SMILES: Cl.CCC(N)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results