CAS RN 773881-43-9
CAS RN 773881-43-9
IUPAC Name:
5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione
Synonyms:
5-octylthieno[3,4-c]pyrrole-4,6-dione
Molecular Weight (g/mol):
265.37
Molecular Formula:
C14H19NO2S
InChi Key:
QMNVUZQWXKLEFP-UHFFFAOYSA-N
SMILES:
CCCCCCCCN1C(=O)C2=CSC=C2C1=O
1
–
1
of
1
results
1
N-n-Octyl-3,4-thiophenedicarboximide 98.0+%, TCI America™
CAS: 773881-43-9 Molecular Formula: C14H19NO2S Molecular Weight (g/mol): 265.37 MDL Number: MFCD23703118 InChI Key: QMNVUZQWXKLEFP-UHFFFAOYSA-N Synonym: 5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione PubChem CID: 67122608 IUPAC Name: 5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione SMILES: CCCCCCCCN1C(=O)C2=CSC=C2C1=O