CAS RN 78-04-6
CAS RN 78-04-6
IUPAC Name:
dibutylstannanebis(ylium) (2E)-but-2-enedioate
Synonyms:
dibutylstannanebis(ylium) fumarate
Molecular Weight (g/mol):
347
Molecular Formula:
C12H20O4Sn
InChi Key:
ZBBLRPRYYSJUCZ-YOTINIEPSA-L
SMILES:
[O-]C(=O)\C=C\C([O-])=O.CCCC[Sn++]CCCC
1
–
1
of
1
results
1
Dibutyltin Maleate (so called) [for PVC stabilizer], TCI America™
CAS: 78-04-6 Molecular Formula: C12H20O4Sn MDL Number: MFCD00014120