CAS RN 83857-96-9
CAS RN 83857-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde 98.0+%, TCI America™
CAS: 83857-96-9 Molecular Formula: C8H11ClN2O Molecular Weight (g/mol): 186.64 MDL Number: MFCD01934396 InChI Key: JLVIHQCWASNXCK-UHFFFAOYSA-N Synonym: 2-butyl-4-chloro-5-formylimidazole,2-butyl-4-chloro-1h-imidazole-5-carbaldehyde,unii-7h170u0swe,2-butyl-5-chloroimidazole-4-carbaldehyde,2-butyl-5-chloro-3h-imidazole-4-carbaldehyde,2-butyl-4 5-chloro-5 4 imidazolecarboxaldehyde,1h-imidazole-4-carboxaldehyde, 2-butyl-5-chloro,2-butyl-4-chloro-1h-imidazole-5-carboxaldehyde,2-butyl-4-chloro-5-formylimidazloe,2-n-butyl-4-chloro-5-formylimidazole PubChem CID: 55176 IUPAC Name: 2-butyl-5-chloro-1H-imidazole-4-carbaldehyde SMILES: CCCCC1=NC(C=O)=C(Cl)N1