accessibility menu, dialog, popup

UserName

CAS RN 85006-25-3

CH3CH3H3COOHNOOOCH3CH3H3C

CAS RN 85006-25-3

IUPAC Name: 2-{[({[(tert-butoxy)carbonyl]amino}oxy)carbonyl]oxy}-2-methylpropane
Synonyms: 2-[({[(tert-butoxycarbonyl)amino]oxy}carbonyl)oxy]-2-methylpropane
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: AGOSGCWATIJZHQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results