accessibility menu, dialog, popup

UserName

CAS RN 88284-48-4

CH3SiH3CCH3OSOOFFF

CAS RN 88284-48-4

IUPAC Name: 2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms: 2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 298.35
Molecular Formula: C10H13F3O3SSi
InChi Key: XBHPFCIWRHJDCP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results