accessibility menu, dialog, popup

UserName

CAS RN 92413-47-3

H3CO

CAS RN 92413-47-3

IUPAC Name: 4'-butyl-[1,1'-bi(cyclohexane)]-4-one
Synonyms: 4'-butyl-[1,1'-bi(cyclohexane)]-4-one
Molecular Weight (g/mol): 236.4
Molecular Formula: C16H28O
InChi Key: LDUYZTOYURUHIX-UHFFFAOYSA-N
SMILES: CCCCC1CCC(CC1)C1CCC(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results