accessibility menu, dialog, popup

UserName

CAS RN 930-55-2

ONN

CAS RN 930-55-2

IUPAC Name: 1-nitrosopyrrolidine
Synonyms: N-nitrosopyrrolidine
Molecular Weight (g/mol): 100.12
Molecular Formula: C4H8N2O
InChi Key: WNYADZVDBIBLJJ-UHFFFAOYSA-N
SMILES: O=NN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results