accessibility menu, dialog, popup

UserName

CAS RN 99328-23-1

CH3OOOOOOOOCH3

CAS RN 99328-23-1

IUPAC Name: 1,4-bis[4-(methoxycarbonyl)phenyl] benzene-1,4-dicarboxylate
Synonyms: 1,4-bis[4-(methoxycarbonyl)phenyl] benzene-1,4-dicarboxylate
Molecular Weight (g/mol): 434.4
Molecular Formula: C24H18O8
InChi Key: XLVVPIVANBPSAY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC(=O)C2=CC=C(C=C2)C(=O)OC2=CC=C(C=C2)C(=O)OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results