missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 98%

CAS: 1515-14-6 | C4H4F6O | 182.07 g/mol

$159.23 - $563.14

Identifiants chimiques

CAS 1515-14-6
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.07
MDL Number MFCD00039259
InChI Key FQDXJYBXPOMIBX-UHFFFAOYSA-N
Synonym hexafluoro-2-methylisopropanol, 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-methyl, hexafluoro-tert-butanol, cf3 2mecoh, 1,1,1,3,3,3-hexafluoro-2-methylisopropanol, methylbistrifluoromethylmethanol, cf3 2c oh ch3, fqdxjybxpomibx-uhfffaoysa, hexafluoro-3-methylisopropanol
PubChem CID 73936
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
SMILES CC(O)(C(F)(F)F)C(F)(F)F
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAL0924506
Afficher les documents
Thermo Scientific Chemicals
L0924506
5 g
chaque for $159.23
Il en reste null
 
AAL0924514
Afficher les documents
Thermo Scientific Chemicals
L0924514
25 g
chaque for $563.14
Il en reste null
 
Description

Description

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

Solubility
Slightly soluble in water.

Notes
Incompatible materials are strong acids, strong bases, strong oxidizing agents, strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Spécifications

Identifiants chimiques

1515-14-6
182.07
FQDXJYBXPOMIBX-UHFFFAOYSA-N
73936
CC(O)(C(F)(F)F)C(F)(F)F
C4H4F6O
MFCD00039259
hexafluoro-2-methylisopropanol, 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-methyl, hexafluoro-tert-butanol, cf3 2mecoh, 1,1,1,3,3,3-hexafluoro-2-methylisopropanol, methylbistrifluoromethylmethanol, cf3 2c oh ch3, fqdxjybxpomibx-uhfffaoysa, hexafluoro-3-methylisopropanol
1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol

Spécifications

1515-14-6
60°C to 62°C
1.295
5 g
hexafluoro-2-methylisopropanol, 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-methyl, hexafluoro-tert-butanol, cf3 2mecoh, 1,1,1,3,3,3-hexafluoro-2-methylisopropanol, methylbistrifluoromethylmethanol, cf3 2c oh ch3, fqdxjybxpomibx-uhfffaoysa, hexafluoro-3-methylisopropanol
FQDXJYBXPOMIBX-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
73936
98%
1.484
C4H4F6O
MFCD00039259
1841110
Slightly soluble in water.
CC(O)(C(F)(F)F)C(F)(F)F
182.07
182.07
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 216-155-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only