missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,1,1,3,3,3-Hexafluoro-2-propanol 99.0+%, TCI America™
Supplier: TCI America H042425G
Specifications
1,1,1,3,3,3-Hexafluoro-2-propanol | |
-4°C | |
58°C | |
MFCD00011651 | |
1760 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
13529 | |
168.04 | |
Liquid |
920-66-1 | |
Colorless | |
C3H2F6O | |
25 g | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
C(C(F)(F)F)(C(F)(F)F)O | |
168.038 | |
CHEBI:63104 | |
≥99.0% (GC) |
Chemical Identifiers
920-66-1 | |
168.038 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
13529 | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
C3H2F6O | |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
CHEBI:63104 | |
C(C(F)(F)F)(C(F)(F)F)O |
Safety and Handling
EINECSNumber : (2)-0291
RTECSNumber : UB6450000
TSCA : Yes