missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoroisopropyl p-Toluenesulfonate 98.0+%, TCI America™
$158.44 - $464.14
Chemical Identifiers
| CAS | 67674-48-0 |
|---|---|
| Molecular Formula | C10H8F6O3S |
| Molecular Weight (g/mol) | 322.22 |
| MDL Number | MFCD00039262 |
| InChI Key | QGSVBFAODCJVIZ-UHFFFAOYSA-N |
| Synonym | 1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester |
| PubChem CID | 98916 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzene-1-sulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F |
Chemical Identifiers
| 67674-48-0 | |
| 322.22 | |
| QGSVBFAODCJVIZ-UHFFFAOYSA-N | |
| 98916 | |
| CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F |
| C10H8F6O3S | |
| MFCD00039262 | |
| 1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzene-1-sulfonate |
Specifications
| 67674-48-0 | |
| White-Yellow | |
| C10H8F6O3S | |
| 5 g | |
| QGSVBFAODCJVIZ-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzene-1-sulfonate | |
| 98916 | |
| ≥98.0% (GC) | |
| 1,1,1,3,3,3-Hexafluoroisopropyl p-Toluenesulfonate |
| 45°C | |
| 134°C | |
| MFCD00039262 | |
| 1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester | |
| CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F | |
| 322.22 | |
| 322.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes