missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane 96.0+%, TCI America™
$106.25 - $238.60
Chemical Identifiers
| CAS | 7422-52-8 |
|---|---|
| Molecular Formula | C13H32O4Si3 |
| Molecular Weight (g/mol) | 336.65 |
| MDL Number | MFCD00053926 |
| InChI Key | YSIQPJVFCSCUMU-UHFFFAOYSA-N |
| PubChem CID | 62635 |
| IUPAC Name | trimethyl-[methyl-[3-(oxiran-2-ylmethoxy)propyl]-trimethylsilyloxysilyl]oxysilane |
| SMILES | C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C |
Chemical Identifiers
| 7422-52-8 | |
| 336.65 | |
| YSIQPJVFCSCUMU-UHFFFAOYSA-N | |
| trimethyl-[methyl-[3-(oxiran-2-ylmethoxy)propyl]-trimethylsilyloxysilyl]oxysilane |
| C13H32O4Si3 | |
| MFCD00053926 | |
| 62635 | |
| C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C |
Specifications
| 7422-52-8 | |
| 98°C | |
| MFCD00053926 | |
| YSIQPJVFCSCUMU-UHFFFAOYSA-N | |
| trimethyl-[methyl-[3-(oxiran-2-ylmethoxy)propyl]-trimethylsilyloxysilyl]oxysilane | |
| 62635 | |
| ≥96.0% (GC) | |
| 1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane |
| Colorless | |
| C13H32O4Si3 | |
| 5 g | |
| C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C | |
| 336.65 | |
| 336.65 | |
| Liquid |
Safety and Handling
TSCA : Yes