Learn More
1,1,1-Trifluoro-2,4-pentanedione, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1468222
Description
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.
Solubility
Soluble in water (slightly) and many organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
1,1,1-Trifluoro-2,4-pentanedione | |
1.274 | |
26°C (79°F) | |
1.389 | |
100 g | |
1705177 | |
Soluble in water (slightly) and many organic solvents. | |
CC(=O)CC(=O)C(F)(F)F | |
154.09 | |
154.09 |
367-57-7 | |
105°C to 107°C | |
C5H5F3O2 | |
MFCD00000427 | |
UN1224 | |
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone | |
SHXHPUAKLCCLDV-UHFFFAOYSA-N | |
1,1,1-trifluoropentane-2,4-dione | |
73943 | |
98% |
Chemical Identifiers
367-57-7 | |
154.09 | |
SHXHPUAKLCCLDV-UHFFFAOYSA-N | |
73943 | |
CC(=O)CC(=O)C(F)(F)F |
C5H5F3O2 | |
MFCD00000427 | |
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone | |
1,1,1-trifluoropentane-2,4-dione |
Safety and Handling
GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 206-698-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only