missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-propanol, 97%

CAS: 374-01-6 | C3H5F3O | 114.067 g/mol

$121.10 - $508.41

Chemical Identifiers

CAS 374-01-6
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.067
MDL Number MFCD00014403
InChI Key GILIYJDBJZWGBG-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
PubChem CID 9774
IUPAC Name 1,1,1-trifluoropropan-2-ol
SMILES CC(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1115006
View Documents
Thermo Scientific Chemicals
L1115006
5 g N/A
Only null left
 
AAL1115014
View Documents
Thermo Scientific Chemicals
L1115014
25 g N/A
Only null left
 
Description

Description

1,1,1-Trifluoro-2-propanol is used as a chemical building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Chemical Identifiers

374-01-6
114.067
GILIYJDBJZWGBG-UHFFFAOYSA-N
9774
CC(C(F)(F)F)O
C3H5F3O
MFCD00014403
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
1,1,1-trifluoropropan-2-ol

Specifications

374-01-6
75°C to 76°C
C3H5F3O
MFCD00014403
UN1987
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
GILIYJDBJZWGBG-UHFFFAOYSA-N
1,1,1-trifluoropropan-2-ol
9774
97%
1.259
25°C (77°F)
1.3175
5 g
1734738
Fully miscible in water.
CC(C(F)(F)F)O
114.067
114.07
1,1,1-Trifluoro-2-propanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 206-773-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only