missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2,2-Tetrachloroethane, 98.5%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 147940025

Catalog No. AC147940025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1,1,2,2-Tetrachloroethane
79-34-5
100.0
147.0°C
98.5%
C2H2Cl4
Cl2CHCHCl2
2.5L
1.58
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
QPFMBZIOSGYJDE-UHFFFAOYSA-N
1,1,2,2-tetrachloroethane
6591
CHEBI:36026
98.5%
98.5%
98.0
1.5800g/mL
Authentic
Glass bottle
1.4930 to 1.4950
MFCD00000848
01, 86
15, 9334
Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
C(C(Cl)Cl)(Cl)Cl
167.85
1.7 mPa.s (28°C)
167.85
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 201-197-8

RTECSNumber : KI8575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only