Learn More
1,1,2,2-Tetrachloroethane, 98.5%, Thermo Scientific Chemicals
$87.83 - $359.17
Chemical Identifiers
CAS | 79-34-5 |
---|---|
MDL Number | MFCD00000848 |
InChI Key | QPFMBZIOSGYJDE-UHFFFAOYSA-N |
Synonym | s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan |
PubChem CID | 6591 |
ChEBI | CHEBI:36026 |
IUPAC Name | 1,1,2,2-tetrachloroethane |
SMILES | C(C(Cl)Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147941000
|
Thermo Scientific Chemicals
147941000 |
100 mL | Glass bottle |
Each for $87.83
|
|
||||
AC147940010
|
Thermo Scientific Chemicals
147940010 |
1 L | Glass bottle |
Each for $359.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-34-5 | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
6591 | |
1,1,2,2-tetrachloroethane |
MFCD00000848 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
CHEBI:36026 | |
C(C(Cl)Cl)(Cl)Cl |
Specifications
79-34-5 | |
100.0 | |
Colorless | |
147.0°C | |
98.5% | |
C2H2Cl4 | |
Cl2CHCHCl2 | |
100 mL | |
1.58 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethane | |
6591 | |
CHEBI:36026 | |
98.5% | |
1,1,2,2-Tetrachloroethane, 98.5% |
98.0 | |
-43.0°C | |
1.5800g/mL | |
Authentic | |
Glass bottle | |
1.4930 to 1.4950 | |
MFCD00000848 | |
01, 86 | |
15, 9334 | |
Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone | |
C(C(Cl)Cl)(Cl)Cl | |
167.85 | |
1.7 mPa.s (28°C) | |
167.85 | |
Liquid |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
GHS Signal Word: Danger
EINECSNumber : 201-197-8
RTECSNumber : KI8575000
TSCA : TSCA
RUO – Research Use Only