missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2-Trichloroethane 98.0+%, TCI America™

$53.32 - $152.08

Chemical Identifiers

CAS 79-00-5
Molecular Formula C2H3Cl3
Molecular Weight (g/mol) 133.40
MDL Number MFCD00000852
InChI Key UBOXGVDOUJQMTN-UHFFFAOYSA-N
Synonym ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
PubChem CID 6574
ChEBI CHEBI:36018
IUPAC Name 1,1,2-trichloroethane
SMILES ClCC(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T038125G
View Documents
TCI America
T038125G
25 g
Each for $53.32
 
T0381500G
View Documents
TCI America
T0381500G
500 g
Each for $152.08
 
Description

Description

Specifications

Chemical Identifiers

79-00-5
133.40
UBOXGVDOUJQMTN-UHFFFAOYSA-N
6574
1,1,2-trichloroethane
C2H3Cl3
MFCD00000852
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
CHEBI:36018
ClCC(Cl)Cl

Specifications

79-00-5
114°C
MFCD00000852
2810
UBOXGVDOUJQMTN-UHFFFAOYSA-N
1,1,2-trichloroethane
6574
133.40
Liquid
Colorless
C2H3Cl3
25 g
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
ClCC(Cl)Cl
133.40
CHEBI:36018
≥98.0% (GC)
1,1,2-Trichloroethane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0055

RTECSNumber : KJ3150000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'