missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trichloropropane 96.0+%, TCI America™

Supplier:  TCI America T06865ML

Catalog No. T06865ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,1,2-Trichloropropane
Colorless
C3H5Cl3
5 mL
2-Chloropropylidene Chloride
CC(C(Cl)Cl)Cl
147.423
147.42
Liquid
598-77-6
133°C
MFCD00018864
1993
GRSQYISVQKPZCW-UHFFFAOYSA-N
1,1,2-trichloropropane
11733
≥96.0% (GC)

Chemical Identifiers

598-77-6
147.423
GRSQYISVQKPZCW-UHFFFAOYSA-N
11733
CC(C(Cl)Cl)Cl
C3H5Cl3
MFCD00018864
2-Chloropropylidene Chloride
1,1,2-trichloropropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0083

RTECSNumber : TZ8925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions