Learn More
1,1':3',1″-Terphenyl-5'-boronic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2764003
Description
A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.
Solubility
Insoluble in water.
Notes
Refrigerated (2-4°C). Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
1,1':3',1''-Terphenyl-5'-boronic acid | |
292°C to 296°C | |
MFCD09953491 | |
3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x | |
MRBZYVMZUBUDAX-UHFFFAOYSA-N | |
(3,5-diphenylphenyl)boronic acid | |
14739363 | |
95% |
128388-54-5 | |
C18H15BO2 | |
1 g | |
Insoluble in water. | |
B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O | |
274.126 | |
274.13 |
Chemical Identifiers
128388-54-5 | |
274.126 | |
MRBZYVMZUBUDAX-UHFFFAOYSA-N | |
14739363 | |
B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O |
C18H15BO2 | |
MFCD09953491 | |
3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x | |
(3,5-diphenylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only