Learn More
1,1,3,3-Tetramethylbutyl isocyanate, 98%, Thermo Scientific Chemicals
$46.04 - $129.77
Chemical Identifiers
CAS | 1611-57-0 |
---|---|
Molecular Formula | C9H17NO |
Molecular Weight (g/mol) | 155.24 |
MDL Number | MFCD00209615 |
InChI Key | XOHBENIMDRFUIH-UHFFFAOYSA-N |
PubChem CID | 407498 |
IUPAC Name | 2-isocyanato-2,4,4-trimethylpentane |
SMILES | CC(C)(C)CC(C)(C)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1870603
|
Thermo Scientific Chemicals
L1870603 |
1 g |
Each for $46.04
|
|
|||||
AAL1870606
|
Thermo Scientific Chemicals
L1870606 |
5 g |
Each for $129.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1611-57-0 | |
155.24 | |
XOHBENIMDRFUIH-UHFFFAOYSA-N | |
2-isocyanato-2,4,4-trimethylpentane |
C9H17NO | |
MFCD00209615 | |
407498 | |
CC(C)(C)CC(C)(C)N=C=O |
Specifications
1611-57-0 | |
170°C to 172°C | |
Pungent | |
1.426 | |
1 g | |
Moisture sensitive | |
XOHBENIMDRFUIH-UHFFFAOYSA-N | |
2-isocyanato-2,4,4-trimethylpentane | |
407498 | |
98% |
0.859 | |
51°C (123°F) | |
C9H17NO | |
MFCD00209615 | |
UN3080 | |
1,1,3,3-tetramethylbutyl isocyanate, tert-octyl isocyanate, 2-isocyanato-2,4,4-trimethyl-pentane, pentane, 2-isocyanato-2,4,4-trimethyl, acmc-1bnst, pentane,2-isocyanato-2,4,4-trimethyl, unavailable-see asdi_catno 500009647, 1,1,3,3-tetramethylbutyl isocyanate∼2,4,4-trimethyl-2-pentyl isocyanate | |
CC(C)(C)CC(C)(C)N=C=O | |
155.24 | |
155.24 | |
1,1,3,3-Tetramethylbutyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H226-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H330-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only