missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1'-Azobis(N,N-dimethylformamide) 97.0+%, TCI America™

$45.43 - $165.90

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
MDL Number MFCD00008318
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A14581G
View Documents
TCI America
A14581G
1 g N/A
 
A14585G
View Documents
TCI America
A14585G
5 g N/A
 
Description

Description

Specifications

Chemical Identifiers

10465-78-8
172.188
VLSDXINSOMDCBK-BQYQJAHWSA-N
5353800
(3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
C6H12N4O2
MFCD00008318
diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
CHEBI:48963
CN(C)C(=O)N=NC(=O)N(C)C

Specifications

10465-78-8
Yellow
MFCD00008318
diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
CN(C)C(=O)N=NC(=O)N(C)C
172.188
CHEBI:48963
≥97.0% (T)
1,1′-Azobis(N,N-dimethylformamide)
112°C
C6H12N4O2
1 g
VLSDXINSOMDCBK-BQYQJAHWSA-N
(3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
5353800
172.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LQ1041000

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
missing translation for 'documents'

missing translation for 'documents'