missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals

$90.53 - $530.02

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
IUPAC Name di(imidazol-1-yl)methanone
SMILES O=C(N1C=CN=C1)N1C=CN=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151810100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
151810100
10 g Glass bottle
Each for $90.53
 
AC151810250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
151810250
25 g Glass Bottle
Each for $181.98
 
AC151811000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
151811000
100 g Glass bottle
Each for $530.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

530-62-1
162.15
PFKFTWBEEFSNDU-UHFFFAOYSA-N
68263
O=C(N1C=CN=C1)N1C=CN=C1
C7H6N4O
MFCD00005286
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
di(imidazol-1-yl)methanone

Specifications

530-62-1
100.0
White to Cream
96% min
C7H6N4O
10 g
01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PFKFTWBEEFSNDU-UHFFFAOYSA-N
di(imidazol-1-yl)methanone
68263
97%
1,1′-Carbonyldiimidazole
96.0
118.0°C to 120.0°C
Authentic
Glass bottle
MFCD00005286
23, V,4, 245
15, 1820
Solubility in water: decomposes. Other solubilities: moderately soluble in thf, soluble in ethanol
O=C(N1C=CN=C1)N1C=CN=C1
162.15
162.15
Crystalline Powder
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-488-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only