missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic acid, 99%

CAS: 5445-51-2 | C6H8O4 | 144.13 g/mol

Supplier:  Thermo Scientific Chemicals 110920050

 View more versions of this product

Catalog No. AC110920050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 1-Cyclobutanedicarboxylic acid
5445-51-2
100.0
White
Authentic
Glass bottle
C4H6(CO2H)2
5 g
1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
C1CC(C1)(C(=O)O)C(=O)O
144.13
CHEBI:35691
99%
Fine Crystalline Powder
99%
98.5
157°C to 161°C
0.1% max. (50°C)
98.5% min. (GC)
C6H8O4
MFCD00001325
09, 724
CCQPAEQGAVNNIA-UHFFFAOYSA-N
cyclobutane-1,1-dicarboxylic acid
2568
144.13
(5 w/v% aq. soln.) clear

Chemical Identifiers

5445-51-2
144.13
CCQPAEQGAVNNIA-UHFFFAOYSA-N
2568
cyclobutane-1,1-dicarboxylic acid
C6H8O4
MFCD00001325
1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
CHEBI:35691
C1CC(C1)(C(=O)O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 226-651-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only