missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil, Thermo Scientific™
Specifications
CAS | 3006-86-8 |
---|---|
Molecular Formula | C14H28O4 |
Molecular Weight (g/mol) | 260.37 |
MDL Number | MFCD00084800 |
InChI Key | HSLFISVKRDQEBY-UHFFFAOYSA-N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC361310100
|
Thermo Scientific Chemicals
361310100 |
10mL | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC361312500
|
Thermo Scientific Chemicals
361312500 |
250mL | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Radical initiatorsSpecifications
3006-86-8 | |
260.37 | |
HSLFISVKRDQEBY-UHFFFAOYSA-N | |
6366 | |
1,1-dichloroethene |
C14H28O4 | |
MFCD00084800 | |
vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene | |
CHEBI:34031 | |
CC(C)(C)OOC1(CCCCC1)OOC(C)(C)C |
RUO â Research Use Only