Learn More
1,1-Dibromo-3,3,3-trifluoroacetone, 95%
CAS: 431-67-4 | C3HBr2F3O | 269.843 g/mol
$154.47 - $575.49
Chemical Identifiers
| CAS | 431-67-4 |
|---|---|
| Molecular Formula | C3HBr2F3O |
| Molecular Weight (g/mol) | 269.843 |
| MDL Number | MFCD00041362 |
| InChI Key | HEPPAPZASXFWTB-UHFFFAOYSA-N |
| Synonym | 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone |
| PubChem CID | 550523 |
| IUPAC Name | 3,3-dibromo-1,1,1-trifluoropropan-2-one |
| SMILES | C(C(=O)C(F)(F)F)(Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAL1082606
|
Thermo Scientific Chemicals
L1082606 |
5 g |
Each for $154.47
|
|
|||||
|
AAL1082614
|
Thermo Scientific Chemicals
L1082614 |
25 g |
Each for $575.49
|
|
|||||
Description
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.
Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.
Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.
Chemical Identifiers
| 431-67-4 | |
| 269.843 | |
| HEPPAPZASXFWTB-UHFFFAOYSA-N | |
| 550523 | |
| C(C(=O)C(F)(F)F)(Br)Br |
| C3HBr2F3O | |
| MFCD00041362 | |
| 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone | |
| 3,3-dibromo-1,1,1-trifluoropropan-2-one |
Specifications
| 431-67-4 | |
| Pungent | |
| 1.4305 | |
| 5 g | |
| 636645 | |
| Soluble in Chloroform. Not miscible or difficult to mix in water. | |
| C(C(=O)C(F)(F)F)(Br)Br | |
| 269.843 | |
| 269.85 | |
| 1,1-Dibromo-3,3,3-trifluoroacetone |
| 111°C to 113°C | |
| C3HBr2F3O | |
| MFCD00041362 | |
| UN3265 | |
| 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone | |
| HEPPAPZASXFWTB-UHFFFAOYSA-N | |
| 3,3-dibromo-1,1,1-trifluoropropan-2-one | |
| 550523 | |
| 95% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only