Learn More
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 113471000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 1-Dichlorodimethyl ether | |
96.0 | |
Colorless to Yellow | |
85.0°C | |
Authentic | |
Glass bottle | |
1.4295 to 1.4325 | |
MFCD00000828 | |
01,220; 02,120; 05,200; 06,171; 07,98; 08,154 | |
methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester | |
COC(Cl)Cl | |
114.96 | |
114.96 | |
Liquid |
4885-02-3 | |
100.0 | |
1.2700g/mL | |
22°C | |
96% min. (GC) | |
C2H4Cl2O | |
Cl2CHOCH3 | |
100 g | |
1.27 | |
GRTGGSXWHGKRSB-UHFFFAOYSA-N | |
methyl 2-sulfanylbenzoate | |
21009 | |
97% |
Chemical Identifiers
4885-02-3 | |
114.96 | |
GRTGGSXWHGKRSB-UHFFFAOYSA-N |
C2H4Cl2O | |
MFCD00000828 | |
COC(Cl)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Harmful if inhaled.
In use,may form flammable/explosive vapor-air mixture.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 225-498-9
RUO – Research Use Only