missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
$80.36 - $219.14
Chemical Identifiers
CAS | 14634-91-4 |
---|---|
Molecular Formula | C36H24FeN6O4S |
Molecular Weight (g/mol) | 692.53 |
MDL Number | MFCD00036428 |
InChI Key | CIWXFRVOSDNDJZ-UHFFFAOYSA-L |
Synonym | ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 |
PubChem CID | 84567 |
SMILES | [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Description
1,10-Phenanthroline iron(II) sulfate is used as an indicator for the iron phenanthroline compound in analytical chemistry. Further, it is used as a redox indicator for making oscillatory Belousov-Zhabotinsky reactions visible.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14634-91-4 | |
692.53 | |
CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
84567 |
C36H24FeN6O4S | |
MFCD00036428 | |
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Specifications
14634-91-4 | |
C36H24FeN6O4S | |
50 g | |
CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
λ²-iron(2+) tris(1,10-phenanthroline) sulfate | |
84567 | |
0.025M aq. soln. | |
1,10-Phenanthroline iron(II) sulfate |
Red | |
MFCD00036428 | |
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
692.53 | |
692.53 | |
Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only