missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Phenanthroline monohydrate, For the Spectrophotometric Determination of Fe, Pd, V, ≥99.0%, MilliporeSigma™ Supelco™

1,10-Phenanthroline monohydrate is a chelating agent.

$105.30 - $415.34

Chemical Identifiers

MDL Number MFCD00149973
Synonym o-Phenanthroline monohydrate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111020158
View Documents
Merck Emd Millipore
775005GF
5 g
Each for $105.30
 
111018525
View Documents
Merck Emd Millipore
7750025GF
25 g
Each for $415.34
 
Description

Description

Specifications

Chemical Identifiers

MFCD00149973
o-Phenanthroline monohydrate

Specifications

100°C to 104°C; 99°C to 104°C
C12H8N2 · H2O
5 g
198.22
5144-89-8
MFCD00149973
o-Phenanthroline monohydrate
≥99.0% (HPLC); ≥99.0% (NT); ≥99.0%
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SF8437000

SDS
missing translation for 'documents'

missing translation for 'documents'