missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ) 95.0+%, TCI America™

$168.38 - $501.73

Chemical Identifiers

CAS 72829-09-5
Molecular Formula C20H34O4
Molecular Weight (g/mol) 338.488
MDL Number MFCD00080499
InChI Key HYQASEVIBPSPMK-UHFFFAOYSA-N
Synonym 1,12-Bis(methacryloyloxy)dodecane
PubChem CID 175191
IUPAC Name 12-(2-methylprop-2-enoyloxy)dodecyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCCCCCCCOC(=O)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D517225G
View Documents
TCI America
D517225G
25 g
Each for $168.38
 
D5172100G
View Documents
TCI America
D5172100G
100 g
Each for $501.73
 
Description

Description

Specifications

Chemical Identifiers

72829-09-5
338.488
HYQASEVIBPSPMK-UHFFFAOYSA-N
175191
CC(=C)C(=O)OCCCCCCCCCCCCOC(=O)C(=C)C
C20H34O4
MFCD00080499
1,12-Bis(methacryloyloxy)dodecane
12-(2-methylprop-2-enoyloxy)dodecyl 2-methylprop-2-enoate

Specifications

72829-09-5
<7
MFCD00080499
1,12-Bis(methacryloyloxy)dodecane
CC(=C)C(=O)OCCCCCCCCCCCCOC(=O)C(=C)C
338.488
338.49
Liquid
Yellow
C20H34O4
25 g
HYQASEVIBPSPMK-UHFFFAOYSA-N
12-(2-methylprop-2-enoyloxy)dodecyl 2-methylprop-2-enoate
175191
≥95.0% (GC)
1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'