missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%

$370.02 - $370.02

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115240250
SDS View Documents
Thermo Scientific Chemicals
115240250
25 g Glass bottle
Each for $370.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4064-06-6
260.28
POORJMIIHXHXAV-UHFFFAOYNA-N
C12H20O6
MFCD00063225

Specifications

4064-06-6
100.0
Yellow
117.0°C (0.1 mmHg)
96% min. (GC)
C12H20O6
MFCD00063225
1.14
-57.5
POORJMIIHXHXAV-UHFFFAOYNA-N
{4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
344833
97%
1, 2:3, 4-Di-O-isopropylidene-D-galactopyranose, 97%
96.0
120.0°C to 122.0°C
1.1400g/mL
Authentic
Glass bottle
1.4668 to 1.4688
25 g
− 57.50 (20.00°C c=3,CHCl3)
n-4-methylphenyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolylacetamide, cbmicro_030789, n-p-tolyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolyl acetamide, nbeta-z-nin-boc-l-beta-homotryptophan, 2,2-diphenyl-n-p-tolyl-acetamide, n-4-methylphenyl-2,2-diphenyl-acetamide, n∼1∼-4-methylphenyl-2,2-diphenylacetamide
CC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
260.28
260.28
Viscous Paste
Safety and Handling

Safety and Handling

EINECSNumber : 223-771-7

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only