missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H32138MD

 View more versions of this product

Catalog No. AAH32138MD


Explore available promotions

Description

Description

1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

Solubility
Soluble in methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

1,2,3,4-Tetrahydro-1-naphthoic acid
82°C to 85°C
MFCD02179127
1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
VDLWTJCSPSUGOA-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
12598146
98%
1914-65-4
C11H12O2
250 mg
Soluble in methanol,and chloroform.
C1CC(C2=CC=CC=C2C1)C(=O)O
176.215
176.22

Chemical Identifiers

1914-65-4
176.215
VDLWTJCSPSUGOA-UHFFFAOYSA-N
12598146
C1CC(C2=CC=CC=C2C1)C(=O)O
C11H12O2
MFCD02179127
1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only