Learn More
1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 175810250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,3,4-Tetrahydro-1-naphthylamine | |
97.5 | |
Colorless to Yellow | |
246.0°C to 247.0°C (714.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5625 to 1.5645 | |
25 g | |
1.02 | |
DETWFIUAXSWCIK-UHFFFAOYNA-N | |
1,2,3,4-tetrahydronaphthalen-1-amine | |
18066 | |
97% |
2217-40-5 | |
100.0 | |
1.0200g/mL | |
>110°C | |
96% min. (GC) | |
C10H14ClN | |
MFCD00001740 | |
12,12 | |
1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine | |
[Cl-].[NH3+]C1CCCC2=CC=CC=C12 | |
183.68 | |
147.22 | |
Liquid |
Chemical Identifiers
2217-40-5 | |
183.68 | |
DETWFIUAXSWCIK-UHFFFAOYNA-N | |
18066 | |
[Cl-].[NH3+]C1CCCC2=CC=CC=C12 |
C10H14ClN | |
MFCD00001740 | |
1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine | |
1,2,3,4-tetrahydronaphthalen-1-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 218-712-7
RUO – Research Use Only