missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H26726AA

 View more versions of this product

Catalog No. AAH26726AA


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline
0.975
>110°C (230°F)
1.5475
5 mL
NRWNXIXJZMSDAU-UHFFFAOYSA-N
2,2,4,7-tetramethyl-3,4-dihydro-1H-quinoline
101035
97%
59388-58-8
123°C (5 mmHg)
C13H19N
MFCD00799587
1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline, 2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-2,2,4,7-tetramethyl, acmc-20ap7g, quinoline,1,2,3,4-tetrahydro-2,2,4,7-tetramethyl
CC1CC(NC2=C1C=CC(=C2)C)(C)C
189.302
189.3

Chemical Identifiers

59388-58-8
189.302
NRWNXIXJZMSDAU-UHFFFAOYSA-N
2,2,4,7-tetramethyl-3,4-dihydro-1H-quinoline
C13H19N
MFCD00799587
101035
CC1CC(NC2=C1C=CC(=C2)C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 261-732-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only