Learn More
1,2,3,4-Tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A149620I
Description
1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air and light sensitive. Heat sensitive. May darken on storage. Stable under recommended storage conditions.
Specifications
1,2,3,4-Tetrahydronaphthalene | |
-36°C | |
206°C to 207°C | |
C10H12 | |
MFCD00001733 | |
UN3082 | |
Air Sensitive | |
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene | |
CXWXQJXEFPUFDZ-UHFFFAOYSA-N | |
1,2,3,4-tetrahydronaphthalene | |
8404 | |
132.21 |
119-64-2 | |
0.966 | |
77°C (171°F) | |
1.54 | |
5000 g | |
1446407 | |
14,9221 | |
It is insoluble in water. | |
C1CCC2=CC=CC=C2C1 | |
132.21 | |
CHEBI:35008 | |
97% |
Chemical Identifiers
119-64-2 | |
132.21 | |
CXWXQJXEFPUFDZ-UHFFFAOYSA-N | |
8404 | |
1,2,3,4-tetrahydronaphthalene |
C10H12 | |
MFCD00001733 | |
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene | |
CHEBI:35008 | |
C1CCC2=CC=CC=C2C1 |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P260-P264b-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 204-340-2
RTECSNumber : QK3850000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only