Learn More
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
$213.44 - $1382.74
Chemical Identifiers
CAS | 119-64-2 |
---|---|
Molecular Formula | C10H12 |
Molecular Weight (g/mol) | 132.21 |
MDL Number | MFCD00001733 |
InChI Key | CXWXQJXEFPUFDZ-UHFFFAOYSA-N |
Synonym | tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene |
PubChem CID | 8404 |
ChEBI | CHEBI:35008 |
IUPAC Name | 1,2,3,4-tetrahydronaphthalene |
SMILES | C1CCC2=CC=CC=C2C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC146730010
|
Thermo Scientific Chemicals
146730010 |
1 L | Glass bottle |
Each for $213.44
|
|
|||||
AC146730025
|
Thermo Scientific Chemicals
146730025 |
2.5 L | Glass bottle |
Each for $420.65
|
|
|||||
AC146730100
|
Thermo Scientific Chemicals
146730100 |
10 L | Metal Drum |
Each for $1,382.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
119-64-2 | |
132.21 | |
CXWXQJXEFPUFDZ-UHFFFAOYSA-N | |
8404 | |
1,2,3,4-tetrahydronaphthalene |
C10H12 | |
MFCD00001733 | |
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene | |
CHEBI:35008 | |
C1CCC2=CC=CC=C2C1 |
Specifications
119-64-2 | |
100.0 | |
0.9730g/mL | |
77°C | |
Glass bottle | |
1.5400 to 1.5420 | |
MFCD00001733 | |
05, IV, 1388 | |
0.973 | |
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene | |
CXWXQJXEFPUFDZ-UHFFFAOYSA-N | |
1,2,3,4-tetrahydronaphthalene | |
8404 | |
CHEBI:35008 | |
98+% | |
1,2,3,4-Tetrahydronaphthalene |
98.0 | |
-35.0°C | |
207.0°C | |
98% min. (GC) | |
C10H12 | |
C10H12 | |
1 L | |
01,1142 | |
15, 9368 | |
Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents | |
C1CCC2=CC=CC=C2C1 | |
132.21 | |
2.2 mPa.s (20°C) | |
132.2 | |
Pure |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May form explosive peroxides.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 204-340-2
RUO – Research Use Only