missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals

$952.26 - $952.26

Chemical Identifiers

CAS 4249-10-9
Molecular Weight (g/mol) 122.21
InChI Key VNPQQEYMXYCAEZ-UHFFFAOYSA-N
Synonym cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile
PubChem CID 77935
SMILES CC1=C(C)C(C)=C(C)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC348070250
SDS View Documents
Thermo Scientific Chemicals
348070250
25 g
Each for $952.26
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4249-10-9
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
77935
122.21
cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile
CC1=C(C)C(C)=C(C)C1

Specifications

4249-10-9
0.8080g/mL
42°C
80% min. (GC)
C9H14
25 g
cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile
CC1=C(C)C(C)=C(C)C1
122.21
122.21
Liquid
Yellow
142.0°C
Authentic
Glass bottle
1.4700 to 1.4760
0.808
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
cyclopentanecarbonitrile
77935
85%
1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only