missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2,3-Benzothiadiazole-5-carbonyl chloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC09402DE
Specifications
1, 2, 3-Benzothiadiazole-5-carbonyl chloride | |
95% | |
C7H3ClN2OS | |
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride | |
C1=CC2=C(C=C1C(=O)Cl)N=NS2 | |
198.624 | |
198.63 |
321309-32-4 | |
Amber Glass Bottle | |
5g | |
VOSTUOLSYXVSND-UHFFFAOYSA-N | |
1,2,3-benzothiadiazole-5-carbonyl chloride | |
2735457 | |
95% |