missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Benzothiadiazole-5-carbonyl chloride, 95%, Thermo Scientific™

$72.88 - $72.88

Chemical Identifiers

CAS 321309-32-4
Molecular Formula C7H3ClN2OS
Molecular Weight (g/mol) 198.624
InChI Key VOSTUOLSYXVSND-UHFFFAOYSA-N
Synonym 1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
PubChem CID 2735457
IUPAC Name 1,2,3-benzothiadiazole-5-carbonyl chloride
SMILES C1=CC2=C(C=C1C(=O)Cl)N=NS2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC09402CB
SDS View Documents
Thermo Scientific Chemicals
CC09402CB
250 mg
Each for $72.88
 
Description

Description

Specifications

Chemical Identifiers

321309-32-4
198.624
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
1,2,3-benzothiadiazole-5-carbonyl chloride
C7H3ClN2OS
VOSTUOLSYXVSND-UHFFFAOYSA-N
2735457
C1=CC2=C(C=C1C(=O)Cl)N=NS2

Specifications

321309-32-4
C7H3ClN2OS
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
C1=CC2=C(C=C1C(=O)Cl)N=NS2
198.624
198.63
1, 2, 3-Benzothiadiazole-5-carbonyl chloride
95%
250 mg
VOSTUOLSYXVSND-UHFFFAOYSA-N
1,2,3-benzothiadiazole-5-carbonyl chloride
2735457
95%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'