Learn More
1,2,3-Thiadiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
$168.55 - $564.29
Chemical Identifiers
CAS | 4100-13-4 |
---|---|
Molecular Formula | C3H2N2O2S |
Molecular Weight (g/mol) | 130.121 |
MDL Number | MFCD00052099 |
InChI Key | HJZYBDPHAHGHAZ-UHFFFAOYSA-N |
Synonym | 1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid |
PubChem CID | 351418 |
IUPAC Name | thiadiazole-4-carboxylic acid |
SMILES | C1=C(N=NS1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2084203
|
Thermo Scientific Chemicals
B2084203 |
1 g |
N/A
|
|
||||||
AAB2084206
|
Thermo Scientific Chemicals
B2084206 |
5 g |
N/A
|
|
||||||
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
4100-13-4 | |
130.121 | |
HJZYBDPHAHGHAZ-UHFFFAOYSA-N | |
351418 | |
C1=C(N=NS1)C(=O)O |
C3H2N2O2S | |
MFCD00052099 | |
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid | |
thiadiazole-4-carboxylic acid |
Specifications
4100-13-4 | |
C3H2N2O2S | |
1 g | |
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid | |
HJZYBDPHAHGHAZ-UHFFFAOYSA-N | |
thiadiazole-4-carboxylic acid | |
351418 | |
97% |
227°C to 228°C | |
MFCD00052099 | |
113707 | |
Slightly soluble in water. | |
C1=C(N=NS1)C(=O)O | |
130.121 | |
130.12 | |
1,2,3-Thiadiazole-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only