missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Trifluoro-5-nitrobenzene, 99%, Thermo Scientific Chemicals

$133.14 - $491.25

Chemical Identifiers

CAS 66684-58-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00456803
InChI Key PTTUMBGORBMNBN-UHFFFAOYSA-N
Synonym 3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
PubChem CID 2782793
IUPAC Name 1,2,3-trifluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(F)C(F)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2030206
View Documents
Thermo Scientific Chemicals
L2030206
5 g
Each for $133.14
 
AAL2030214
View Documents
Thermo Scientific Chemicals
L2030214
25 g
Each for $491.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66684-58-0
177.08
PTTUMBGORBMNBN-UHFFFAOYSA-N
2782793
[O-][N+](=O)C1=CC(F)=C(F)C(F)=C1
C6H2F3NO2
MFCD00456803
3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
1,2,3-trifluoro-5-nitrobenzene

Specifications

66684-58-0
75°C (167°F)
1.486
5 g
2455018
PTTUMBGORBMNBN-UHFFFAOYSA-N
1,2,3-trifluoro-5-nitrobenzene
2782793
99%
1.52
C6H2F3NO2
MFCD00456803
UN2810
3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
[O-][N+](=O)C1=CC(F)=C(F)C(F)=C1
177.08
177.08
1,2,3-Trifluoro-5-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 266-447-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only