Learn More
1,2,4,5-Tetrabromobenzene, 94%
CAS: 636-28-2 | C6H2Br4 | 393.70 g/mol
$63.38 - $197.71
Chemical Identifiers
| CAS | 636-28-2 |
|---|---|
| Molecular Formula | C6H2Br4 |
| Molecular Weight (g/mol) | 393.70 |
| MDL Number | MFCD00000063 |
| InChI Key | QCKHVNQHBOGZER-UHFFFAOYSA-N |
| Synonym | benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo |
| PubChem CID | 12486 |
| IUPAC Name | 1,2,4,5-tetrabromobenzene |
| SMILES | BrC1=CC(Br)=C(Br)C=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1353909
|
Thermo Scientific Chemicals
L1353909 |
10 g |
Each for $63.38
|
|
|||||
|
AAL1353918
|
Thermo Scientific Chemicals
L1353918 |
50 g |
Each for $197.71
|
|
|||||
Description
1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
| 636-28-2 | |
| 393.70 | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 12486 | |
| BrC1=CC(Br)=C(Br)C=C1Br |
| C6H2Br4 | |
| MFCD00000063 | |
| benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo | |
| 1,2,4,5-tetrabromobenzene |
Specifications
| 636-28-2 | |
| C6H2Br4 | |
| 10 g | |
| benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 1,2,4,5-tetrabromobenzene | |
| 12486 | |
| 94% |
| 175°C to 180°C | |
| MFCD00000063 | |
| 1365830 | |
| Insoluble in water. | |
| BrC1=CC(Br)=C(Br)C=C1Br | |
| 393.70 | |
| 393.7 | |
| 1,2,4,5-Tetrabromobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-253-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only